Orders LinkContactUsLinkComponent
SymLine® Automatic Switching for Container
SymLine® Automatic Switching for Container
Non-ATEX set: automatic switching for container - 1× built-in Signalbox2, EU/UK/US, 2-channel, 2 switch inputs and 2 switch outputs for peripheral units (SCTV106548), 1× SwitchBox: control unit for SCAT built-in signal box or signal box T1/T5 (SCTV106730), 1× 3-way ball valve, 3× GL 25 (m), electric, material: stainless steel (SCTV160178), 1× 3-way ball valve mount, stainless steel (SCTV106580), 2× signal cable for electronic level monitoring, length = 3 m (590-0739)
Catalog # SCTV106751
Supplier:   S.C.A.T.
SymLine® Automatic Switching for Container
Catalog # SCTV106751
Supplier:  S.C.A.T.
SymLine® Automatic Switching for Container
SymLine® Automatic Switching for Container
Non-ATEX set: automatic switching for container - 1× built-in Signalbox2, EU/UK/US, 2-channel, 2 switch inputs and 2 switch outputs for peripheral units (SCTV106548), 1× SwitchBox: control unit for SCAT built-in signal box or signal box T1/T5 (SCTV106730), 1× 3-way ball valve, 3× GL 25 (m), electric, material: stainless steel (SCTV160178), 1× 3-way ball valve mount, stainless steel (SCTV106580), 2× signal cable for electronic level monitoring, length = 3 m (590-0739)
Price
1 617,56 €
Log in for availability
1 * 1 ST
Product Details & Documents
The ready-to-use automatic switching sets prevent the overflowing of disposal containers and the associated leakage of hazardous, toxic, or explosive liquid waste.
  • Coordinated system for monitoring the safe filling of disposal containers
  • For connection to two disposal containers (not included in the set)
  • Set consists of Built-in Signalbox2, SwitchBox, ball valve, and two signal cables
  • Optionally available as an ATEX version with an additional switch amplifier for two channels
Expandable sets for safe fill level monitoring during disposal in containers.
Specifications
  • Description:
    Non-ATEX set: automatic switching for container - 1× built-in Signalbox2, EU/UK/US, 2-channel, 2 switch inputs and 2 switch outputs for peripheral units (SCTV106548), 1× SwitchBox: control unit for SCAT built-in signal box or signal box T1/T5 (SCTV106730), 1× 3-way ball valve, 3× GL 25 (m), electric, material: stainless steel (SCTV160178), 1× 3-way ball valve mount, stainless steel (SCTV106580), 2× signal cable for electronic level monitoring, length = 3 m (590-0739)
  • Seq:
    1