CD138 MONOCLONAL ANTIBODY 1 * 100 µl
Catalog # ENZOENZABS6830100
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Deep well plates and cover mats
Supplier: Brand
Cooling systems, CoolSafe™
Supplier: DIVERSIFIED BIOTECH
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Cryoboxes, coloured, Nalgene®
Supplier: Thermo Fisher Scientific
Containers, wide mouth, with screw caps
Supplier: KARTELL
Transfer pipettes, Samco™ Fine Tip
Supplier: SAMCO
Scoops, with bent handle, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Tubes Cryo.S™ 4 and 5 ml
Supplier: Greiner Bio-One
Measuring scoops, PP
Supplier: SEMADENI
Sampling scoops, long handle, disposable, LaboPlast®/SteriPlast®
Supplier: BURKLE