Orders LinkContactUsLinkComponent
Membrane potential detection kit, MITO-ID®
Membrane potential detection kit, MITO-ID®
MITO-ID® membrane potential detection kit, 100 Tests
Catalog # ENZOENZ51018K100
Supplier:   Enzo Life Sciences
Membrane potential detection kit, MITO-ID®
Catalog # ENZOENZ51018K100
Supplier:  Enzo Life Sciences
CAS Number:  
Membrane potential detection kit, MITO-ID®
Membrane potential detection kit, MITO-ID®
MITO-ID® membrane potential detection kit, 100 Tests
CAS Number:  
Price
674,00 €
Log in for availability
1 * 100 Tests
Product Details & Documents
MITO-ID® membrane potential detection kit monitors energetic status using a simple mix-and-read, no-wash protocol. It measures mitochondrial membrane potential with a cationic dye that fluoresces either green or orange depending upon membrane potential status.
  • Highly sensitive detection of mitochondrial membrane potential decline
  • Aids in the drug research process in preclinical drug safety assessment (ADME-Tox)
  • Suitable for chemical/environmental toxicity screening and characterisation
  • True mix-and-read homogeneous assay for live cells
  • Optimized for the measurement of mitochondria membrane potential (MMP) and cell viability by fluorescence microscopy and flow cytometry
Cell-based assays for evaluating the functional status of mitochondria are useful tools for elucidating the role of mitochondrial activity in drug-induced toxicity, the apoptosis cascade and other cellular and biochemical processes.
Delivery information: Kit contains MITO-ID® MP detection reagent, necrosis detection reagent, CCCP control, 10X assay buffer 1 and 50X assay buffer 2.
Specifications
Specifications
  • Pk:
    100 Tests
  • Description:
    MITO-ID® membrane potential detection kit
Specifications