Orders LinkContactUsLinkComponent
Membrane potential detection kit, MITO-ID® | Avantor
Membrane potential detection kit, MITO-ID®
MITO-ID® Membrane potential detection kit, 25 Tests
Catalog # ENZOENZ510180025
Supplier:   Enzo Life Sciences
Membrane potential detection kit, MITO-ID®
Catalog # ENZOENZ510180025
Supplier:  Enzo Life Sciences
CAS Number:  
Membrane potential detection kit, MITO-ID®
Membrane potential detection kit, MITO-ID®
MITO-ID® Membrane potential detection kit, 25 Tests
CAS Number:  
Price
244,00 €
Log in for availability
1 * 25 Tests
Product Details & Documents
MITO-ID® membrane potential detection kit monitors energetic status using a simple mix-and-read, no-wash protocol. It measures mitochondrial membrane potential with a cationic dye that fluoresces either green or orange depending upon membrane potential status.
  • Highly sensitive detection of mitochondrial membrane potential decline
  • Aids in the drug research process in preclinical drug safety assessment (ADME-Tox)
  • Suitable for chemical/environmental toxicity screening and characterisation
  • True mix-and-read homogeneous assay for live cells
  • Optimized for the measurement of mitochondria membrane potential (MMP) and cell viability by fluorescence microscopy and flow cytometry
Cell-based assays for evaluating the functional status of mitochondria are useful tools for elucidating the role of mitochondrial activity in drug-induced toxicity, the apoptosis cascade and other cellular and biochemical processes.
Lieferumfang: Kit contains MITO-ID® MP detection reagent, necrosis detection reagent, CCCP control, 10X assay buffer 1 and 50X assay buffer 2.
Specifications
Specifications
  • Pk:
    25 Tests
  • Description:
    MITO-ID® Membrane potential detection kit
Specifications
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.