Order Entry
Austria
Orders LinkContactUsLinkComponent
Human recombinant Activin-A

Human recombinant Activin-A

10 µG
Catalog # ENZOALX2018090010
Supplier:   Enzo Life Sciences
Human recombinant Activin-A
Catalog # ENZOALX2018090010
Supplier:  Enzo Life Sciences
CAS Number:  
Human recombinant Activin-A

10 µG

CAS Number:  
Price
356,00 €
Log in for availability
1 * 10 µG
Product Details & Documents

Produced in E. coli.

  • Carrier-free

Non-glycosylated homodimer, containing two 117 amino acid chains.

Specifications

Specifications

  • Pk:
    10 µG
  • Cat. No.:
    ENZOALX2018090010
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Activin-A
  • Formulation:
    Lyophilized from a sterile filtered solution containing 0,1% TFA.
  • Protein synonyms:
    Inhibin beta-1|FRP|FSH-releasing protein
  • Purity:
    >95% (HPLC, Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm)
  • Species:
    Human
  • Biological activity:
    The activity is determined by the ability to induce cytoxicity of MPC-11 cells which is typically observed at concentations <2 ng/ml.
  • Molecular weight:
    ~26,2kDa

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.