Orders LinkContactUsLinkComponent
Diagonal cutters
Diagonal cutters
Diagonal cutter, chrome plated, handles insulated with multi-component grips
Catalog # KNIP7005125
Supplier:   KNIPEX WERKE
Diagonal cutters
Catalog # KNIP7005125
Supplier:  KNIPEX WERKE
Diagonal cutters
Diagonal cutters
Diagonal cutter, chrome plated, handles insulated with multi-component grips
Price
33,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Vanadium electric steel; forged, oil-hardened. The indispensable diagonal cutter for all-round use. Chrome plated head. With two-coloured insulated multi-component grips. Insulated upto 1000 V.
  • High quality material and precise workmanship for long service life
  • Precision cutting edges for soft and hard wire
  • Cutting edges additionally induction hardened
Certifications: DIN ISO 5749. VDE-tested.
Specifications
  • Description:
    Diagonal cutter, chrome plated, handles insulated with multi-component grips
  • Seq:
    01