1 COMPONENT C8H5O4K 60MG/L IN H2O 1 * 1 L
Catalog # CMPLRM0484171L
Supplier: CUSTOM MADE CHEMICALS LAB
1 COMPONENT C8H5O4K 60MG/L IN H2O 1 * 1 L
Catalog # CMPLRM0484171L
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
Ga 20mg/l In 10mg/l Ir 10mg/l Sc 200mg/l Tm 10mg/l Rh 10mg/l 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
NEOMYCIN ELISA KIT 1 * 1 KIT
Supplier: ENZO LIFE SCIENCES
Ammonia solution ≥25% in water, TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Supplier: Honeywell Chemicals
Potassium hydroxide ≥85% ACS (max. 2% K₂CO₃)
Supplier: Thermo Fisher Scientific
VARIO TOTAL CHLORINE DPD 25 ML 1 * 100 items
Supplier: Lovibond