BUFFER PH2 EXP FOR VWR HITACHI AAA - Buffer without Ethanol - Ethanol has to been added before usage 1 * 1 L
Catalog # HPLCB16001EXP
Supplier: COLLECTION PREFIX HPLC
BUFFER PH2 EXP FOR VWR HITACHI AAA - Buffer without Ethanol - Ethanol has to been added before usage 1 * 1 L
Catalog # HPLCB16001EXP
Supplier: COLLECTION PREFIX HPLC
Frequently Bought Together
pH/mV/°C meters, handheld, Portavo® 902 and 904/904 ATEX
Supplier: KNICK
Short-term tubes for accuro®
Supplier: Dräger
Gas detection tubes for ammonia
Supplier: Dräger
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR®, Accessories for Pocket Colorimeters, MCP 100 and MCT 100
Supplier: VWR Collection
VWR® HCO 304, Conductivity Meter, Handheld
Supplier: VWR Collection
SmartBlocks for ThermoMixer® C and ThermoStat C
Supplier: EPPENDORF
Accessories for cell counter, EVE™
Supplier: NANOENTEK
Accessories for MAS-100®
Supplier: Merck
Accessories for VWR® V-1200, UV-1600PC, UV-3100PC
Supplier: COLLECTION PREFIX HPLC
Accessories for titrators
Supplier: Mettler - Toledo
VWR®, Single-Use Heads for Air Samplers
Supplier: VWR Collection
OIL IMPACTOR 0.1-1.0 MG/M3 1 * 10 items
Supplier: Dräger
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
Titration beakers
Supplier: Mettler - Toledo
Accessory for airflow indicator
Supplier: Dräger
FlashGel™ DNA system
Supplier: LONZA
Titrator, Titronic® 300
Supplier: SI Analytics
Plastic cuvette for LICO
Supplier: Hach