Order Entry
Austria
Orders LinkContactUsLinkComponent
Cooling elements and frames

Cooling elements and frames

Cooling elements, 37 °C, Tempshell, Red, 10...40 °C, 275×155×60 mm
210×110×30 mm
Catalog # DLTTA100041
Supplier:   DELTA T
Cooling elements and frames
Catalog # DLTTA100041
Supplier:  DELTA T
Cooling elements and frames

Cooling elements, 37 °C, Tempshell, Red, 10...40 °C, 275×155×60 mm
210×110×30 mm

Price
108,00 €
Log in for availability
1 * 1 PAAR
Product Details & Documents

Molecular medical elements for different temperature handling of samples ranges from −32 °C to +38 °C.

  • Made of PE and natural in colour
  • Sealing made of laminated aluminium film with a protected cap
  • A liquid crystal thermometer in the elements shows its current temperature

Used for transportation and storage of samples like blood products, homograft’s, cell cultures, stem cells, antibodies, proteins and other temperature sensitive medications.

Inside dimension: 210×110×30 mm

Specifications
  • Temperature Range:
    10...40 °C
  • Colour:
    Red
  • Description:
    Cooling elements, 37 °C, Tempshell
  • Dimensions:
    275×155×60 mm
    210×110×30 mm
  • Seq:
    06