Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Fluorite Objectives for Revolve Microscope
Plan fluorite, 0,13 NA aperture, 17 mm
Catalog # ECHLCMAG0042
Supplier:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Catalog # ECHLCMAG0042
Supplier:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Plan fluorite, 0,13 NA aperture, 17 mm
Price
556,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Documents
Specifications
  • Magnification:
  • Description:
    Plan fluorite, 0,13 NA aperture
  • Seq:
    27
  • Working distance:
    17 mm