BUFFER PH 9.180 AT 25 C ISO 17034 1 * 500 mL
Catalog # CMPLPH219.L5
Supplier: CUSTOM MADE CHEMICALS LAB
BUFFER PH 9.180 AT 25 C ISO 17034 1 * 500 mL
Catalog # CMPLPH219.L5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
N-Nitroso-ethyl-isopropylamine 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
Indium Standard for ICP 1 * 100 mL
Supplier: Merck
Simazine(2-chloro-4,6-bis(ethylamino)-1,3,5-triazine) ≥99.5%
Supplier: VWR Chemicals
SOLUTION–NEXION DET LIM KED BLANK 1 * 1 items
Supplier: PerkinElmer
4-Methoxyphenol 1 * 100 mg
Supplier: EHRENSTORFER
Chinomethionat certified 1 * 0,25 g
Supplier: EHRENSTORFER
Pharmaceutical analytical standard, Promazine-d6 hydrochloride, reference material, Honeywell Fluka™
Supplier: Honeywell Chemicals
1,4-Dioxane
Supplier: EHRENSTORFER
Isobenzan certified 1 * 10 mg
Supplier: EHRENSTORFER
Perindopril Related Compounds, Reference Standard
Supplier: USP
Total organic carbon (TOC) standards
Supplier: REAGECON
TopFluor Oleic Acid 1 * 1 Each
Supplier: Merck