Order Entry
Austria
Orders LinkContactUsLinkComponent
Digital pH Sensor, Memosens CPS71E

Digital pH Sensor, Memosens CPS71E

Digital pH sensor Memosens CPS71E, 0,8 - 14 bar (abs), ATEX II 1G Ex ia IIC T3/T4/T6 Ga, 0 - 14 pH, 0...140 °C, 120 mm
Catalog # ENDRCPS71E-BA7BTB2
Supplier:   Endress+Hauser
Digital pH Sensor, Memosens CPS71E
Catalog # ENDRCPS71E-BA7BTB2
Supplier:  Endress+Hauser
Digital pH Sensor, Memosens CPS71E
Digital pH Sensor, Memosens CPS71E

Digital pH sensor Memosens CPS71E, 0,8 - 14 bar (abs), ATEX II 1G Ex ia IIC T3/T4/T6 Ga, 0 - 14 pH, 0...140 °C, 120 mm

Price
664,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

With its contamination-resistant reference system, Memosens CPS71E is a digital pH sensor for highly contaminated or toxic media such as strong acids or bases.

  • The special ion trap of the electrode prevents poisoning of the junction and the reference, offers resistance to strong acids and bases and ensures a longer service life of the sensor
  • The optional pressurized reference ensures reliable measurements in blocking media
  • Optional upside-down mounting for flexible installation
  • Inductive cable connection and non-contact signal transmission improve process integrity by preventing problems with moisture or corrosion
  • On-site sensor replacement minimizes process downtime and costs

Memosens 2.0 technology enables trend identification, predictive maintenance and advanced IIoT services with extended data storage for calibration and process information.

Bestellinformation: For optional items or more information, contact your Avantor representative.

Lieferumfang: Digital pH sensor Memosens CPS71E.

Specifications
  • Description:
    Digital pH sensor Memosens CPS71E, 0,8 - 14 bar (abs), ATEX II 1G Ex ia IIC T3/T4/T6 Ga
  • Length:
    120 mm
  • Seq:
    03
  • Temp.:
    0...140 °C
  • pH:
    0 - 14 pH
Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.