ContactUsLinkComponent
,, ,,1 component: Metolachlor ESA sodium salt [CAS:947601-85-6] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store in a freezer at -18C or below,, 1 * 1 mL
Catalog # CMPLRD0025151
,, ,,1 component: Metolachlor ESA sodium salt [CAS:947601-85-6] 100mg/l in Acetonitrile ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 months.,, ,,Store in a freezer at -18C or below,, 1 * 1 mL
Catalog # CMPLRD0025151
Frequently Bought Together