Order Entry
Austria
Orders LinkContactUsLinkComponent
Assistent® Brushes for Flasks

Assistent® Brushes for Flasks

Natural bristle, —, 70 mm, Wood, 440 mm
Catalog # HECH41912010
Assistent® Brushes for Flasks
Catalog # HECH41912010
Assistent® Brushes for Flasks
Assistent® Brushes for Flasks

Natural bristle, —, 70 mm, Wood, 440 mm

Price
24,60 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Straight horsehair brushes with galvanized iron thread handle.

Spezifikationen
  • Bristle material:
    Natural bristle
  • Brush head Ø:
    70 mm
  • Handle material:
    Wood
  • Seq:
    05
  • Total length:
    440 mm
  • Brush head length:
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.