ContactUsLinkComponent
9 components: 1000mg/l each of 4-tert-Octylphenol Monoethoxylate [CAS:2315-67-5] 4-tert-Octylphenol Diethoxylate [CAS:2315-61-9] 4-Nonylphenol-mono-ethoxylate [CAS:104-35-8] 4-Nonylphenol-di-ethoxylate [CAS:20427-84-3] Polyethylene Glycol Mon 1 * 5 Ampoul
Catalog # CMPLRD0737001.5
9 components: 1000mg/l each of 4-tert-Octylphenol Monoethoxylate [CAS:2315-67-5] 4-tert-Octylphenol Diethoxylate [CAS:2315-61-9] 4-Nonylphenol-mono-ethoxylate [CAS:104-35-8] 4-Nonylphenol-di-ethoxylate [CAS:20427-84-3] Polyethylene Glycol Mon 1 * 5 Ampoul
Catalog # CMPLRD0737001.5
Frequently Bought Together