Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Microscope slides

Slides, cut edges, soda-lime glass, White, 1 mm, 75×75 mm
Catalog # MARI1120250
Supplier:   MARIENFELD
Microscope slides
Catalog # MARI1120250
Supplier:  MARIENFELD
Microscope slides
Microscope slides

Slides, cut edges, soda-lime glass, White, 1 mm, 75×75 mm

Price
882,71 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Slides designed for in vitro diagnostic (IVD) applications.

  • Pre-washed and ready to use
  • Either ground or cut versions
  • Soda-lime glass, Hydrolytic Class 3

Extra-white slides are used for low auto-fluorescence applications.

Documents

Certificates
Search by Lot Number
Specifications
  • Colour:
    White
  • Description:
    Slides, cut edges, soda-lime glass
  • L×W:
    75×75 mm
  • Seq:
    06
  • Thickness:
    1 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.