Order Entry
Austria
Orders LinkContactUsLinkComponent
QuickConnect™ Mop Handles

QuickConnect™ Mop Handles

QuickConnect™ carbon fiber telescopic handle, 175 - 848 cm (69 - 334")
Catalog # CONTCFH334
Supplier:   CONTEC
QuickConnect™ Mop Handles
Catalog # CONTCFH334
Supplier:  CONTEC
QuickConnect™ Mop Handles
QuickConnect™ Mop Handles

QuickConnect™ carbon fiber telescopic handle, 175 - 848 cm (69 - 334")

Price
733,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Interchangeable mop handles, available in carbon fiber, anodised aluminum, stainless steel, or fiberglass.

  • Compatible with all QuickConnect™ frames
  • Multiple lengths available

Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles.

Telescoping handles are available in lightweight carbon fiber, as well as anodised aluminum, durable stainless steel, or fiberglass. Choose from a variety of lengths.

Specifications
  • Description:
    QuickConnect™ carbon fiber telescopic handle
  • Length:
    175 - 848 cm (69 - 334")
  • Seq:
    05