ESHMUNO A 1 * 100 mL
Catalog # 1.20089.0100_T
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
N,N-Dimethylformamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
Supplier: Merck
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
1,6-Dibromohexane for synthesis, Sigma-Aldrich®
Supplier: Merck
Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent, VOLUSOL®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
8 components: 100mg/l each of Al Cr Cu Fe Mn Ba Zn Zr in HNO3 5% 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
Petroleum spirit, 35…60 °C ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals