MANGANESE CHL. TETRAHYDRATE + NOT. CHANG 1 * 125 g
Catalog # SPCMYY2351-125GM
Supplier: Spectrum Chemical
MANGANESE CHL. TETRAHYDRATE + NOT. CHANG 1 * 125 g
Catalog # SPCMYY2351-125GM
Supplier: Spectrum Chemical
Frequently Bought Together
Sodium formate ≥99%, AnalaR NORMAPUR®
Supplier: VWR Chemicals
Sodium hydrogen carbonate 99.7-100.3%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Aluminium oxide, γ-phase
Supplier: Merck
di-Potassium hydrogen phosphate ≥99.0% analytical reagent
Supplier: VWR Chemicals
Potassium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Water, EMSURE® for analysis, Supelco®
Supplier: Merck
Tin(II) chloride dihydrate ≥98%, AnalaR NORMAPUR® analytical reagent (max. 0.05 ppm Hg)
Supplier: VWR Chemicals