Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TPE PURPLE CAP 1 CS = 10000 PACK 1 * 10.000 items
Catalog # JETBFTD100020
Supplier:
GUANGZHOU JET BIOFILTRATION
Expand image
TPE PURPLE CAP 1 CS = 10000 PACK 1 * 10.000 items
Catalog # JETBFTD100020
Supplier:
GUANGZHOU JET BIOFILTRATION
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
VWR® Disposable Spatulas, PP
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection