ContactUsLinkComponent
This product is a kit for quantifying genomic DNA from E. coli by qPCR (probe method). It can be used for residual DNA testing for b iopharmaceuticals and their manufacturing processes. 1 * 100 Tests
Catalog # WAKF290-85301
This product is a kit for quantifying genomic DNA from E. coli by qPCR (probe method). It can be used for residual DNA testing for b iopharmaceuticals and their manufacturing processes. 1 * 100 Tests
Catalog # WAKF290-85301
Frequently Bought Together