ContactUsLinkComponent
This product is a kit for quantifying genomic DNA from CHO cells by qPCR (probe method). It can be used for residual DNA testing for antibody pharmaceuticals and their manufacturing processes. 1 * 100 Tests
Catalog # WAKF294-85201
This product is a kit for quantifying genomic DNA from CHO cells by qPCR (probe method). It can be used for residual DNA testing for antibody pharmaceuticals and their manufacturing processes. 1 * 100 Tests
Catalog # WAKF294-85201
Frequently Bought Together