BOTTOM PLATE COMPLETE FOR MAS-100 NT 1 * 1 items
Catalog # MBVA101.219
Supplier: MBV
Frequently Bought Together
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Accessories for PIPETBOY acu / pro
Supplier: Integra
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Cover gasket 1 * 1 items
Supplier: VWR Collection
Accessories for HandyStep® electronic
Supplier: Brand
Lithium Polymer Rechargable Battery
Supplier: EPPENDORF
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Accessories for Transferpette® electronic pipettes
Supplier: Brand
Accessories for Dispensette®
Supplier: Brand
Accessories for accu-jet® pro
Supplier: Brand
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
Rotors and accessories for Megafuge® 8 centrifuges
Supplier: Thermo Scientific
Accessories for water purification millipore
Supplier: Merck
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
Accessories for CO2 Incubators, Heracell VIOS
Supplier: Thermo Scientific