ContactUsLinkComponent
4 bouteilles de 125 mL 2 components: 10mg/l each of Fe Ca in HCl 2% CRM, ISO 17034 and ISO 17025 Conservation 12 months. (6 months after opening) Ambient Temp. 1 * 4 Bottle
Catalog # CMPLRM103164L125.4
4 bouteilles de 125 mL 2 components: 10mg/l each of Fe Ca in HCl 2% CRM, ISO 17034 and ISO 17025 Conservation 12 months. (6 months after opening) Ambient Temp. 1 * 4 Bottle
Catalog # CMPLRM103164L125.4
Frequently Bought Together