DS BUFFER 1L 1 * 1 L
Catalog # OMEGDS-1000
Supplier: OMEGA BIO-TEK
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
ClipTip™ Pipette Tips
Supplier: Thermo Fisher Scientific
SLX-MLUS BUFFER 1 * 1 L
Supplier: OMEGA BIO-TEK
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
PerfeCTa® qPCR ToughMix®
Supplier: Quantabio