Orders LinkContactUsLinkComponent
Wire Baskets for Test Tubes
Wire Baskets for Test Tubes
140×140×140 mm
Catalog # LINK51108211414
Wire Baskets for Test Tubes
Catalog # LINK51108211414
Wire Baskets for Test Tubes
140×140×140 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Wire basket of welded mesh w 8 mm, d 1 mm for test tubes and more. Made up of stainless steel W-no. 1.4301, cube shape, electrochemically polished.
Specifications
  • Seq:
    03
  • W×D×H:
    140×140×140 mm