ContactUsLinkComponent
DNA Isolation from Blood This 384-prep system kit isolates and purifies genomic DNA (gDNA) from whole blood. The kit uses SPRI paramagnetic bead-based technology to effectively produce a high recovery of high quality DNA for downstream applications s 1 * 384 items
Catalog # BECKC34881
Supplier:  Beckman Coulter
DNA Isolation from Blood This 384-prep system kit isolates and purifies genomic DNA (gDNA) from whole blood. The kit uses SPRI paramagnetic bead-based technology to effectively produce a high recovery of high quality DNA for downstream applications s 1 * 384 items
Catalog # BECKC34881
Supplier:  Beckman Coulter
Frequently Bought Together