Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
5 COMPONENTS STANDARD IN WATER 250 ML 1 * 250 mL
Catalog # CMPLRM102703L25
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
5 COMPONENTS STANDARD IN WATER 250 ML 1 * 250 mL
Catalog # CMPLRM102703L25
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
ALKALINE-CYANIDE REAGENT 1 * 500 mL
Supplier: Hach
Acid red 52, Technical Grade, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Potassium hydroxide 45% in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Bromophenol blue
Supplier: Hach
Direct Red 23, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Acid red 27 ≥85-95% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Potassium hydroxide 8 mol/l (8 N) in aqueous solution
Supplier: Hach
Reagents for hydrazine determination using Lovibond® MD photometers
Supplier: Lovibond
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY