Orders LinkContactUsLinkComponent
Codesoft Lite Software
Codesoft Lite Software
Codesoft lite prepetual online 3 year SMA
Catalog # BRDY362916
Supplier:   Brady
Codesoft Lite Software
Catalog # BRDY362916
Supplier:  Brady
Codesoft Lite Software
Codesoft lite prepetual online 3 year SMA
Price
914,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Codesoft Lite label design software can be used for asset tracking, barcode labelling, and inventory and inspection labelling applications.
  • Only available as a single user license
  • Limited to 1 thermal printer
Languages availble include Arabic (Egypt), Czech, Danish, Dutch, English, Finnish, French, German, Hebrew, Hungarian, Italian, Norwegian (Bokmål), Polish, Portuguese, Romanian, Russian, Spanish, Swedish, and Turkish.
Compatible with BBP11, BBP12, BBP72, PR Plus, IP Series, PAM3000, PAM3600, THT 76mm Core, and TLS-PC LINK printers.
Specifications
  • Description:
    Codesoft lite prepetual online 3 year SMA
  • Seq:
    04