SODIUM ACETATE ANHYDROUS USP 1 * 500 g
SPCMSO104-500GM
Frequently Bought Together
Sodium chloride ≥99%, GPR RECTAPUR®
: VWR Chemicals
Potassium iodide ≥99%, GPR RECTAPUR®
: VWR Chemicals
Ammonium metavanadate ≥99.0% analytical reagent
: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals