ContactUsLinkComponent
SYRINGE FILTER 0.45 UM PES 25MMX1.17IN EZFlow® Syringe Filter, Sample Prep, PES, Foxx Life Sciences 1 * 100 ST
Catalog # VWRMNP381-3216-OEM
Supplier:  VWR Products
SYRINGE FILTER 0.45 UM PES 25MMX1.17IN EZFlow® Syringe Filter, Sample Prep, PES, Foxx Life Sciences 1 * 100 ST
Catalog # VWRMNP381-3216-OEM
Supplier:  VWR Products
Frequently Bought Together