ContactUsLinkComponent
GAM Agar was developed by the Gifu University Medical School (Japan) to isolate and culture anaerobic bacteria from clinical samples. It can also be used for antibiotic sensitivity tests (other than sulphonamide drugs). 1 * 1 items
Catalog # NISS05420-GAM-0300
GAM Agar was developed by the Gifu University Medical School (Japan) to isolate and culture anaerobic bacteria from clinical samples. It can also be used for antibiotic sensitivity tests (other than sulphonamide drugs). 1 * 1 items
Catalog # NISS05420-GAM-0300
Frequently Bought Together