BUFFER PHM 1 * 50 mL
OMEGPHM-50
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
: VWR Collection
Decontamination solution, RNase AWAY®
: Thermo Fisher Scientific
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Single-use syringes, 3-piece, HENKE-JECT®
: HENKE SASS WOLF