RASAL3 ANTIBODY - C-TERMINAL REGION 1 * 100 µl
Catalog # AVIVARP79758_P050
Supplier: Aviva Systems Biology
RASAL3 ANTIBODY - C-TERMINAL REGION 1 * 100 µl
Catalog # AVIVARP79758_P050
Supplier: Aviva Systems Biology
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Supplier: Corning
Transfer pipettes
Supplier: SARSTEDT
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION
Stomacher Model 400 Eco Bag - TVC. Dimensions 155 x 200 mm 1 * 500 items
Supplier: Seward (FermionX)
Microplates, 96-well
Supplier: Greiner Bio-One
384-well clear polystyrene microplates, Corning®
Supplier: Corning
Coloured cap inserts, Capinsert™
Supplier: Simport Scientific
Stoppers, flange plug caps
Supplier: Simport Scientific
Sampling scoops, LaboPlast Bio / SteriPlast Bio
Supplier: BURKLE
Multiwell cell culture plates, CELLSTAR®
Supplier: Greiner Bio-One
Racks for culture tubes, stainless steel
Supplier: BELLCO GLASS