ContactUsLinkComponent
Agilent Tapestation 2200 compatible robotic tips | 30 uL vol., filter, sterile | 384 format | 50 racks/case | Compare to part number : 5067-5153 1 * 19.200 items
Catalog # 736-1083
Supplier:  FLUOTICS
Agilent Tapestation 2200 compatible robotic tips | 30 uL vol., filter, sterile | 384 format | 50 racks/case | Compare to part number : 5067-5153 1 * 19.200 items
Catalog # 736-1083
Supplier:  FLUOTICS
Frequently Bought Together