NRS Neutralising Rinse Solution, Volume 1 mL 1 * 500 items
MEDWMW786
Frequently Bought Together
VWR®, Igientest HACCP
: VWR Collection
BAT Broth and BAT Agar
: DOEHLER GROUP
VWR® Scholten's Dehydrated Culture Medium
: VWR Chemicals
Culture media, ready to use, in plates
: MERCK LIFE SCIENCE
VWR® Culture Media, Ready To Use, In Tubes
: VWR Chemicals
VWR® Ready to Use Media Plates (90 mm), Irradiated
: VWR Chemicals
Nutrient pad sets
: Dr. Möller & Schmelz
MAC CONKEY AGAR 90MM PLATE 1 * 20 items
: LIOFILCHEM
Accessories for Anaerobic Systems, Oxoid
: THERMO FISHER DIAGNOSTICS
LB broth
: Thermo Fisher Scientific
Sodium hydrogen selenite
: THERMO FISHER DIAGNOSTICS
Culti-Loops™ <i>Staphylococcus aureus</i> subsp. <i>aureus</i> ATCC™ 6538P™
: THERMO FISHER DIAGNOSTICS