PHENOLPHTHALEIN 1% W/V IN PYRIDINE 1 * 100 mL
Catalog # CMPLFC846197
Supplier: CUSTOM MADE CHEMICALS LAB
PHENOLPHTHALEIN 1% W/V IN PYRIDINE 1 * 100 mL
Catalog # CMPLFC846197
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Fine dosage syringes, Omnifix®-F Solo
Supplier: B. Braun
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Imidazole ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor