GLYCERIN NATURAL USP 1 * 500 mL
SPCMG1589-500MLPL
Frequently Bought Together
2-Propanol, EMPROVE® EXPERT Ph. Eur., BP, USP for preparative chromatography, SAFC®
: MERCK PRODUCTION CHEMICALS
Hexane (mixture of isomers) ≥98.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Xylene (mixture of isomers) 99% ACS
: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
Sodium hexanitritocobaltate(III)
: Thermo Fisher Scientific
Tetrahydrofuran 99.8% unstabilised for HPLC
: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Tetrachloroethylene ≥99%
: Thermo Fisher Scientific
Methanol ≥99.6%, Laboratory Reagent
: Honeywell Chemicals
N,N-Dimethylacetamide 99.5+% for analysis
: Thermo Fisher Scientific