DEXTROSE ANHYDROUS USP EP BP JP BIOCERT. 1 * 12 kg
Catalog # SPCMD9350-12KG
Supplier: Spectrum Chemical
DEXTROSE ANHYDROUS USP EP BP JP BIOCERT. 1 * 12 kg
Catalog # SPCMD9350-12KG
Supplier: Spectrum Chemical
Frequently Bought Together
Benzoic acid methyl ester for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
o-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
3-(Chloromethyl)benzoyl chloride 98%
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99%
Supplier: Thermo Fisher Scientific
(±)-2-Butanol ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 99+% for analysis
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Diethyl ether, anhydrous ≥99% stabilised ACS
Supplier: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific