1 component: Chloroform [CAS:67-66-3] 2000mg/l in Methanol 1 * 5 Ampoul
Catalog # CMPLRD0089211.5
Supplier: CUSTOM MADE CHEMICALS LAB
1 component: Chloroform [CAS:67-66-3] 2000mg/l in Methanol 1 * 5 Ampoul
Catalog # CMPLRD0089211.5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Cuvettes and tubes for Picco, Multy, Nova and Prove
Supplier: Merck
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals