Tetrachloroethene [CAS:127-18-4] 2000ug/ml in Methanol 1 * 5 Ampoul
Catalog # CMPLP253029.5
Supplier: CUSTOM MADE CHEMICALS LAB
Tetrachloroethene [CAS:127-18-4] 2000ug/ml in Methanol 1 * 5 Ampoul
Catalog # CMPLP253029.5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
EOSIN RAL 555 1L 1 * 1 L
Supplier: RAL
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Test kits, manganese, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide ≥98%, pellets, EMPROVE® ESSENTIAL Ph. Eur., BP, FCC, JP, NF, E524, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sulphate test strips, Dosatest, VWR®
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
BrightVision+ Anti-IgG Antibody (Poly-HRP)
Supplier: IMMUNOLOGIC
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach