DrySyn OCTO MINI PLUS Complete Kit includes 3x DrySyn OCTO MINI Reaction Stations, 48x shortened tubes, 200x caps, 200x septa, 30x stirrer bars and 1x DrySyn MULTI-M base. 1 * 1 KIT
Catalog # ASYNADS19MINIPLUS
Supplier: ASYNT
Frequently Bought Together