Contains 10 components: 10ug/ml each of Sb Au Hf Ir Pd Pt Rh Ru Te Sn in HCl 10% HNO3 1% ,, 1 * 100 mL
CMPL8500-6948.L1
Contains 10 components: 10ug/ml each of Sb Au Hf Ir Pd Pt Rh Ru Te Sn in HCl 10% HNO3 1% ,, 1 * 100 mL
CMPL8500-6948.L1
Frequently Bought Together
Kjeldahl tablets, Kjeltabs
: THOMPSON & CAPPER
Test strips, peracetic acid, Dosatest
: VWR Chemicals
pH indicator test strips, pH-Fix
: MACHEREY-NAGEL
Test papers, lead acetate papers
: MACHEREY-NAGEL
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
: VWR Chemicals
NANOCOLOR® Photometric Tube Tests, Nitrite
: MACHEREY-NAGEL