Closing bracket adjustable CB/E7 1 * 1 items
Catalog # BIND6006-0736
Supplier: Binder
Frequently Bought Together
Millex® Syringe Filters, PTFE Membrane
Supplier: Merck
Sacrifical body 1 * 1 items
Supplier: VWR Collection
Overpressur plug 1 * 1 items
Supplier: VWR Collection
NEW ORING 1000 SET- SINGLE (ALL) 1 * 10 items
Supplier: VWR Collection
Glass wool 30g 1 * 1 items
Supplier: BUCHI
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach
Tip ejector for EHP200,EHP250(LF) pipette 1 * 1 items
Supplier: VWR Collection
CONDENSER V-700/710 LEGACY SET 1 * 1 SET
Supplier: BUCHI
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
ignition contact 1 * 1 items
Supplier: IKA
FRONT PNL TOUCH SCREEN 1 * 1 items
Supplier: VWR Collection
Accessories for Transferpette® S pipettes
Supplier: Brand
PCB RCT BASIC 230V 1 * 1 items
Supplier: IKA