SIMPLECHECK DENSITY BROMOBENZENE 1.495 1 * 15 mL
Catalog # METN30466987
Supplier: Mettler - Toledo
Frequently Bought Together
Diphenylamine 1 g/l in sulphuric acid R Reag. Ph. Eur. 1032101
Supplier: VWR Chemicals
Conductivity standards, Certipur® Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Supplier: VWR Chemicals
pH buffer, single-use, SINGLET™
Supplier: Hach
pH buffer solutions
Supplier: Hach
pH Standard Solutions, IUPAC, Certified
Supplier: Hach