ContactUsLinkComponent
Panthera Tec pol Cloud Binocular Head Siedentopf type, 25º Inclined With Built-in Digital Camera cmos 1/1.8 Still Resolution 8.0mp (3840x2160)Live Resolution Full HD (1920x1080)HDMI, Ethernet And WI-FI Widefield uc-wf10x/20m 1 * 1 items
Catalog # MOTI1100403300241
Supplier:  MOTIC
undefined
Panthera Tec pol Cloud Binocular Head Siedentopf type, 25º Inclined With Built-in Digital Camera cmos 1/1.8 Still Resolution 8.0mp (3840x2160)Live Resolution Full HD (1920x1080)HDMI, Ethernet And WI-FI Widefield uc-wf10x/20m 1 * 1 items
Catalog # MOTI1100403300241
Supplier:  MOTIC
Frequently Bought Together