ContactUsLinkComponent
SpectraMax Fluorescence Polarization (FP) (fluorescein) Detection Cartridge collects all the necessary data in a single read of a mi croplate. Compatible with SpectraMax PARADIGM and/or SpectraMax i3/i3X systems. The resulting speed and performance E 1 * 1 items
Catalog # MLDV0200-7009-TR
Supplier:  Molecular Devices
SpectraMax Fluorescence Polarization (FP) (fluorescein) Detection Cartridge collects all the necessary data in a single read of a mi croplate. Compatible with SpectraMax PARADIGM and/or SpectraMax i3/i3X systems. The resulting speed and performance E 1 * 1 items
Catalog # MLDV0200-7009-TR
Supplier:  Molecular Devices
Frequently Bought Together