Lochdeckel Edelstahl Kantenschutz 300x0.6mm 1 * 1 items
Catalog # MBVA101.292
Supplier: MBV
Frequently Bought Together
Hygiene monitoring system, HY-LiTE® 2
Supplier: Merck
OIL IMPACTOR 0.1-1.0 MG/M3 1 * 10 items
Supplier: Dräger
VWR® Counter-Pen™ Cell Counter
Supplier: VWR Collection
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
Handheld reflectometer, Reflectoquant® RQflex® 20
Supplier: Merck
VWR®, Accessories for Pocket Colorimeters, MCP 100 and MCT 100
Supplier: VWR Collection
Tubes and systems for active sampling
Supplier: Dräger
VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters
Supplier: VWR Collection
Nessler cylinders
Supplier: witeg Labortechnik
Accessories for titrators
Supplier: Mettler - Toledo
VWR® pHenomenal® pH 1100L/1100LB pH/mV/°C Meter, Bench
Supplier: VWR Collection
VWR® PV4 Visible Spectrophotometer
Supplier: VWR Collection
Luer-to-MicroTight® adapter, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Density meters, Densito and DensitoPro
Supplier: Mettler - Toledo
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
Accessories for buchi melting point apparatus M560 and M565
Supplier: BUCHI
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Ubbelohde viscometers, uncalibrated
Supplier: SI Analytics