Order Entry
Austria
Orders LinkContactUsLinkComponent
Accessories for Toggle Drums

Accessories for Toggle Drums

Drum lid, 316L SS, 470 mm int.Ø of drum
Catalog # SAMPA429-470
Supplier:   Sampling Systems
Accessories for Toggle Drums
Catalog # SAMPA429-470
Supplier:  Sampling Systems
Accessories for Toggle Drums

Drum lid, 316L SS, 470 mm int.Ø of drum

Price
405,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Drum lid, 316L SS, 470 mm int.Ø of drum
  • Seq:
    02