Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HF 0.1% 100mg/l each of Ge Mo Nb Sb Sn Ta W Zr Ti 1 * 100 mL
Catalog # CMPLRM095034L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
HF 0.1% 100mg/l each of Ge Mo Nb Sb Sn Ta W Zr Ti 1 * 100 mL
Catalog # CMPLRM095034L1
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
Supplier: Merck
Potassium hydrogen phthalate, Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for alkalimetry, Supelco®
Supplier: Merck
Buffer solutions in sachets
Supplier: Mettler - Toledo
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck